Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Flonoltinib TFA | 2928093-29-0 | 98.0% | C27H35F4N7O3 | 10MG
SDP

Catalog No. 5000260135
Encompass_Preferred

Flonoltinib TFA is the trifluoroacetate salt of a potent, orally active small-molecule inhibitor of JAK2 and FLT3 kinases, used for preclinical oncology research. It exhibits low-nanomolar potency, induces apoptosis and cell-cycle arrest in cancer cell lines, and has shown antitumor activity in animal models.

  • Low-nanomolar potency against JAK2 and FLT3 (JAK2 IC50 0.7 nM; FLT3 IC50 4 nM).
  • Activity against JAK1 and JAK3 (IC50 26 nM and 39 nM), useful for kinase profiling.
  • Supplied as the TFA salt, solid, yellow to orange, suitable for formulation studies.
  • High reported purity (~98%), supporting reproducible biological testing.
  • Water solubility reported (33.33 mg/mL; may require ultrasonic), aiding assay preparation.
  • Storage recommendations: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • For laboratory research use only; not for human therapeutic use.

Catalog No. 50-002-60135 Supplier Medchemexpress LLC Supplier No. HY130247A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Flonoltinib TFA is the trifluoroacetate salt of a potent, orally active small-molecule inhibitor of JAK2 and FLT3 kinases, used for preclinical oncology research. It exhibits low-nanomolar potency, induces apoptosis and cell-cycle arrest in cancer cell lines, and has shown antitumor activity in animal models.

  • Low-nanomolar potency against JAK2 and FLT3 (JAK2 IC50 0.7 nM; FLT3 IC50 4 nM).
  • Activity against JAK1 and JAK3 (IC50 26 nM and 39 nM), useful for kinase profiling.
  • Supplied as the TFA salt, solid, yellow to orange, suitable for formulation studies.
  • High reported purity (~98%), supporting reproducible biological testing.
  • Water solubility reported (33.33 mg/mL; may require ultrasonic), aiding assay preparation.
  • Storage recommendations: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • For laboratory research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.