Learn More
Medchemexpress LLC Flonoltinib TFA | 2928093-29-0 | 98.04% | 25 MG
SDP
Catalog No. 5000341603
Encompass_Preferred
Flonoltinib TFA is a potent and orally active dual JAK2/FLT3 inhibitor, demonstrating anti-cancer activity. It effectively inhibits JAK2, FLT3, JAK1, and JAK3, and has shown significant effects in both laboratory and animal studies.
  • Potent dual JAK2/FLT3 inhibitor
  • Exhibits anti-cancer activity
  • Down-regulates p-FLT3 in a dose-dependent manner
  • Induces apoptosis and G1/G0 cell cycle arrest in MV4-11 cells
  • Shows significant antitumor effects in NOD/SCID mouse models
Catalog No. 50-003-41603 Supplier Medchemexpress LLC Supplier No. 25MGHY130247A
Flonoltinib TFA is a potent and orally active dual JAK2/FLT3 inhibitor, demonstrating anti-cancer activity. It effectively inhibits JAK2, FLT3, JAK1, and JAK3, and has shown significant effects in both laboratory and animal studies.
  • Potent dual JAK2/FLT3 inhibitor
  • Exhibits anti-cancer activity
  • Down-regulates p-FLT3 in a dose-dependent manner
  • Induces apoptosis and G1/G0 cell cycle arrest in MV4-11 cells
  • Shows significant antitumor effects in NOD/SCID mouse models