Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Floor & Wall Protector Shield™
SDP

Catalog No. 19044050 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-044-050 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-044-050 Supplier GOJO™ Supplier No. 4121WHT18
  • $164.00
    $118.65 / Case of 1

    Save $45.35

    28% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Floor and wall protector Shield™ for ES™ and CS™ dispensers.

  • Floor & Wall Protector Shield™ for ES™ and CS™ Dispensers
  • Attaches to bottom of dispenser
  • Combined drip tray and splash guard to protect floors and wall during typical use
TRUSTED_SUSTAINABILITY

Specifications

Product Type Floor and Wall Protector
Color White
Dimensions (L x W x H) 3.86 x 4.68 x 5.98 in.
Holds Hand Sanitizer
Material ABS
Mounting Wall Mount
No. of Compartments 1
Product Style Front-loading with Opening
For Use With (Application) Floor and Wall Drip and Splash Protection
For Use With (Equipment) GOJO ES and CS Dispensers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.