Learn More
MSC Vikan Broom Head, 15 1/4 in, Medium Bristle, Polyester, Green
Catalog No. 502838884
Encompass
This broom head is designed for effective cleaning in demanding environments. Constructed from durable materials, it features a unique bristle system that minimizes contamination risk and bristle loss, making it ideal for sweeping both dry and wet particles in high-risk areas. Its design supports color-coding systems for enhanced hygiene protocols.
  • Durable polypropylene construction with medium polyester bristles.
  • Angled filament security units minimize contamination and bristle loss.
  • Suitable for sweeping dry and wet surfaces in high-risk areas.
  • Color-coded design helps reduce cross-contamination.
  • Acid and chemical resistant.
  • Features a European threaded handle connection.
  • Certified for food contact applications.
Catalog No. 50-283-8884 Supplier MSC Supplier No. 54686548
This broom head is designed for effective cleaning in demanding environments. Constructed from durable materials, it features a unique bristle system that minimizes contamination risk and bristle loss, making it ideal for sweeping both dry and wet particles in high-risk areas. Its design supports color-coding systems for enhanced hygiene protocols.
  • Durable polypropylene construction with medium polyester bristles.
  • Angled filament security units minimize contamination and bristle loss.
  • Suitable for sweeping dry and wet surfaces in high-risk areas.
  • Color-coded design helps reduce cross-contamination.
  • Acid and chemical resistant.
  • Features a European threaded handle connection.
  • Certified for food contact applications.