Learn More
MSC Scot Laboratories Floor Finish Stripper, No-Rinse, Concentrated, 5 gal
Catalog No. 502836450
Encompass
This chemically active floor finish stripper quickly dissolves all water-based floor finishes. Its no-rinse formula reduces the lengthy time typically needed for floor stripping. The process involves mopping it on, waiting a few minutes, agitating, and then removing the liquefied finish with a squeegee-equipped wet vac. Any residual stripper film on the floor will evaporate completely.
  • No-rinse formula
  • Leaves no residue
  • Easy to use: mop on, vac off
  • Industrial strength
Catalog No. 50-283-6450 Supplier MSC Supplier No. 36152395
This chemically active floor finish stripper quickly dissolves all water-based floor finishes. Its no-rinse formula reduces the lengthy time typically needed for floor stripping. The process involves mopping it on, waiting a few minutes, agitating, and then removing the liquefied finish with a squeegee-equipped wet vac. Any residual stripper film on the floor will evaporate completely.
  • No-rinse formula
  • Leaves no residue
  • Easy to use: mop on, vac off
  • Industrial strength