Learn More
Benchmark Products Inc Benchmark Suite Short Loop Floor Mop, High-Performance Ultra-Microfibers, ISO Class 5 Compatible, Unit: each
SDP
Catalog No. NC1597640
Encompass
The BENCHMARK PRODUCTS INC Suite Short Loop Floor Mop provides unparalleled absorbency for critical cleaning environments. Made from high-performance ultra-microfibers, it's ideal for achieving ISO Class 5 compliance. This mop effectively removes liquids, spills, and contaminants while generating low particles, ensuring a cleaner, safer workspace. Packaged conveniently with 5 mops per pack and a case quantity of 25, it stands out for its exceptional performance and effectiveness against various contaminants.
Catalog No. NC1597640 Supplier Benchmark Products Inc Supplier No. YMFY1825
The BENCHMARK PRODUCTS INC Suite Short Loop Floor Mop provides unparalleled absorbency for critical cleaning environments. Made from high-performance ultra-microfibers, it's ideal for achieving ISO Class 5 compliance. This mop effectively removes liquids, spills, and contaminants while generating low particles, ensuring a cleaner, safer workspace. Packaged conveniently with 5 mops per pack and a case quantity of 25, it stands out for its exceptional performance and effectiveness against various contaminants.