Learn More
MSC Karcher Floor Nozzle, Aluminium, 370 mm, DN 40, 1 ea
Catalog No. 502816294
Encompass
This aluminium floor nozzle is designed for high-performance industrial vacuum cleaners. It offers efficient debris collection with a robust construction suitable for demanding applications.
  • Constructed from durable aluminum
  • Features a 370 mm length for extensive cleaning coverage
  • Equipped with a DN 40 standard nominal width connection
  • Compatible with specific IVC, IVM, and IVR-L series industrial vacuum machines
Catalog No. 50-281-6294 Supplier MSC Supplier No. 16616435
This aluminium floor nozzle is designed for high-performance industrial vacuum cleaners. It offers efficient debris collection with a robust construction suitable for demanding applications.
  • Constructed from durable aluminum
  • Features a 370 mm length for extensive cleaning coverage
  • Equipped with a DN 40 standard nominal width connection
  • Compatible with specific IVC, IVM, and IVR-L series industrial vacuum machines