Learn More
Grainger Adam Equipment Floor Scale, Steel, 2,500 lb Capacity, 48 in. x 48 in. Surface
Catalog No. 502794389
Encompass_Preferred
This floor platform scale is designed for demanding industrial environments, featuring a rugged diamond-plated steel construction for added grip and durability. The platform accommodates heavy loads of up to 2,500 lb, making it ideal for pallet weighing and loading dock applications. This digital scale platform is supplied without an indicator, providing the flexibility to integrate with a compatible indicator to best suit specific workflow requirements.
  • Capacity of 2,500 lb
  • Diamond-plated steel surface provides added grip
  • Rugged construction suitable for industrial use
  • Supplied with 15 ft. shielded cable
  • Compatible with a variety of indicators for system customization
  • Certified to CE, FCC, and UL standards
Catalog No. 50-279-4389 Supplier Grainger Supplier No. 19YN50
This floor platform scale is designed for demanding industrial environments, featuring a rugged diamond-plated steel construction for added grip and durability. The platform accommodates heavy loads of up to 2,500 lb, making it ideal for pallet weighing and loading dock applications. This digital scale platform is supplied without an indicator, providing the flexibility to integrate with a compatible indicator to best suit specific workflow requirements.
  • Capacity of 2,500 lb
  • Diamond-plated steel surface provides added grip
  • Rugged construction suitable for industrial use
  • Supplied with 15 ft. shielded cable
  • Compatible with a variety of indicators for system customization
  • Certified to CE, FCC, and UL standards