Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC ZEP Floor Sealer Polish, 55 gal, Mild Scent, 1 ea

Catalog No. 502819933
Encompass

This ZEP Solo Sealer and Polish is a milky white liquid formulated for comprehensive floor care. It is designed to seal and polish floors, providing both protection and an enhanced finish.

  • Formulated for floor care applications
  • Milky white liquid with a slight, mild, ammoniacal odor
  • Supplied in a 55 gal drum
  • Soluble in both cold and hot water
  • PH range of 8-9
  • Density of 1.02 g/cm3

Catalog No. 50-281-9933 Supplier MSC Supplier No. 17690264
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This ZEP Solo Sealer and Polish is a milky white liquid formulated for comprehensive floor care. It is designed to seal and polish floors, providing both protection and an enhanced finish.

  • Formulated for floor care applications
  • Milky white liquid with a slight, mild, ammoniacal odor
  • Supplied in a 55 gal drum
  • Soluble in both cold and hot water
  • PH range of 8-9
  • Density of 1.02 g/cm3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.