Learn More
Accuform™ Floor Sign - PLEASE WAIT HERE UNTIL CALLED UPON (Footprint Symbol)
Shop All AccuformNMC ProductsDescription
- Constructed of thick adhesive vinyl
- Subsurface printed and laminated to protect the graphics and message from heavy pedestrian traffic
- Non-slip, pebble-textured surface provides added durability
- Strong, permanent, acrylic adhesive back and peel-off liner
- Adheres firmly to smooth, clean, dry surfaces
Specifications
Specifications
| Mounting Type | Self-adhesive |
| Languages | English |
| Dimensions (L x W) | 3 x 36 in. |
| Displayed Text | PLEASE WAI HERE UNTIL CALLED UPON |
| Color | White |