Learn More
Grainger Unger AquaDozer Floor Squeegee, Single-Blade, 18 in., 1 ea.
Catalog No. 502789801
Encompass_Preferred
This floor squeegee is designed for efficient removal of water, mud, debris, and slush from smooth, even surfaces. Featuring a single-blade design without pockets or folds, it helps prevent bacterial growth, making it suitable for environments requiring high sanitation standards. The head is constructed with a durable 14-gauge powder-coated steel frame and a premium EPDM rubber blade for heavy-duty performance.
  • Features an 18 in. wide straight blade
  • Constructed with a durable 14-gauge powder-coated steel frame
  • Utilizes a high-quality EPDM rubber blade
  • Includes a reinforced zinc alloy clamp-style socket for threaded handles
  • Designed for industrial use on surfaces like asphalt and concrete
Catalog No. 50-278-9801 Supplier Grainger Supplier No. 169T26
This floor squeegee is designed for efficient removal of water, mud, debris, and slush from smooth, even surfaces. Featuring a single-blade design without pockets or folds, it helps prevent bacterial growth, making it suitable for environments requiring high sanitation standards. The head is constructed with a durable 14-gauge powder-coated steel frame and a premium EPDM rubber blade for heavy-duty performance.
  • Features an 18 in. wide straight blade
  • Constructed with a durable 14-gauge powder-coated steel frame
  • Utilizes a high-quality EPDM rubber blade
  • Includes a reinforced zinc alloy clamp-style socket for threaded handles
  • Designed for industrial use on surfaces like asphalt and concrete