Learn More
Kuhner Shaker Inc FLOOR STAND 1XISF1-X 765MM W
Catalog No. 502570057
Encompass
Floor stand, 1xISF1-X, 765mm with castors ..the floor stand is mounted on an adapter plate with 4 lifting castors..Limits:..200RPM (50mm)..250RPM (25mm)..300RPM (12.5mm)
Catalog No. 50-257-0057 Supplier Kuhner Shaker Inc Supplier No. SMX1561A
Add to Cart
Add to Cart
Request a Quote
Floor stand, 1xISF1-X, 765mm with castors ..the floor stand is mounted on an adapter plate with 4 lifting castors..Limits:..200RPM (50mm)..250RPM (25mm)..300RPM (12.5mm)