Learn More
Kuhner Shaker Inc FLOOR STAND 2XISF1-X 400MM W
Catalog No. 502570058
Encompass
Floor stand, 2xISF1-X, 400mm with castors the floor stand is mounted on an adapter plate with 4 lifting castors..Limits:..200RPM (50mm)..250RPM (25mm)..300RPM (12.5mm)
Catalog No. 50-257-0058 Supplier Kuhner Shaker Inc Supplier No. SMX1560A
Add to Cart
Add to Cart
Request a Quote
Floor stand, 2xISF1-X, 400mm with castors the floor stand is mounted on an adapter plate with 4 lifting castors..Limits:..200RPM (50mm)..250RPM (25mm)..300RPM (12.5mm)