Learn More
Kuhner Shaker Inc FLOOR STAND FOR STACKING UP TO
Catalog No. 502570304
Encompass
Floor Stand for stacking up to two ISF-1W or X Series Kuhner Shaker Incubators, 400mm high.
Catalog No. 50-257-0304 Supplier Kuhner Shaker Inc Supplier No. SM1560
Floor Stand for stacking up to two ISF-1W or X Series Kuhner Shaker Incubators, 400mm high.