Learn More
MSC 3M Stripper Pad, Black, Coarse, Nylon/Polyester, 20 in Diameter
Catalog No. 502839652
Encompass
This stripping pad is designed to effectively remove old, worn-out floor finishes and sealers, and to clean heavily soiled surfaces like concrete when used with appropriate chemicals. It features a durable construction of nylon and polyester fibers with abrasive particles, ensuring consistent high performance and can be washed and reused multiple times to reduce costs.
  • Removes old, soiled floor finishes and sealers.
  • Cleans heavily soiled surfaces such as concrete.
  • Washable and reusable, helping to reduce supply costs.
  • Creates a smooth, refined surface ready for finishing.
  • Constructed from durable nylon and polyester fibers with abrasive particles.
  • Maintains consistent high performance throughout its life.
  • Resistant to water, detergents, and typical floor maintenance cleaners.
  • Green Seal certified for environmental innovation, promoting extended useful life.
Catalog No. 50-283-9652 Supplier MSC Supplier No. 89464648
This stripping pad is designed to effectively remove old, worn-out floor finishes and sealers, and to clean heavily soiled surfaces like concrete when used with appropriate chemicals. It features a durable construction of nylon and polyester fibers with abrasive particles, ensuring consistent high performance and can be washed and reused multiple times to reduce costs.
  • Removes old, soiled floor finishes and sealers.
  • Cleans heavily soiled surfaces such as concrete.
  • Washable and reusable, helping to reduce supply costs.
  • Creates a smooth, refined surface ready for finishing.
  • Constructed from durable nylon and polyester fibers with abrasive particles.
  • Maintains consistent high performance throughout its life.
  • Resistant to water, detergents, and typical floor maintenance cleaners.
  • Green Seal certified for environmental innovation, promoting extended useful life.