Learn More
Grainger Floor Tape Blue 1 1/2 inx1 in T PK545
Catalog No. 502749111
Encompass_Preferred
The Brady Floor Marking Tape T-14D462 is a 2" wide yellow and black striped vinyl tape used to mark floors, aisles, and walkways. It has an aggressive adhesive that adheres to most floor surfaces including concrete, tile, wood, etc. The tape is highly visible and slip resistant. It can withstand foot and vehicle traffic. Rolls are 2" x 36 yards long. The tape meets OSHA standards for floor marking in facilities. It has excellent durability and adhesion.
Catalog No. 50-274-9111 Supplier Grainger Supplier No. 14D462
The Brady Floor Marking Tape T-14D462 is a 2" wide yellow and black striped vinyl tape used to mark floors, aisles, and walkways. It has an aggressive adhesive that adheres to most floor surfaces including concrete, tile, wood, etc. The tape is highly visible and slip resistant. It can withstand foot and vehicle traffic. Rolls are 2" x 36 yards long. The tape meets OSHA standards for floor marking in facilities. It has excellent durability and adhesion.