Learn More
Grainger Floor Tape Yellow 1 3/32 inx1 in L PK750
Catalog No. 502749083
Encompass_Preferred
The Grainger Approved Floor Tape is a 2" wide pressure-sensitive vinyl tape designed for marking floors, aisles, and traffic lanes. It features a high-tack adhesive that adheres to most common floor surfaces including concrete, tile, and wood. The tape has a thickness of 6 mils and is available in five colors: yellow, white, red, green, and black. It can withstand foot and light vehicle traffic and is resistant to moisture, abrasion, and chemicals. This versatile floor marking tape provides a neat, clearly visible line for directing traffic, outlining work areas, and promoting safety in warehouses, factories, schools, and other facilities.
Catalog No. 50-274-9083 Supplier Grainger Supplier No. 14D476
The Grainger Approved Floor Tape is a 2" wide pressure-sensitive vinyl tape designed for marking floors, aisles, and traffic lanes. It features a high-tack adhesive that adheres to most common floor surfaces including concrete, tile, and wood. The tape has a thickness of 6 mils and is available in five colors: yellow, white, red, green, and black. It can withstand foot and light vehicle traffic and is resistant to moisture, abrasion, and chemicals. This versatile floor marking tape provides a neat, clearly visible line for directing traffic, outlining work areas, and promoting safety in warehouses, factories, schools, and other facilities.