Learn More
Medchemexpress LLC HY-100562 100mg Medchemexpress, Flopropione CAS:2295-58-1 Purity:>98%
SDP
Catalog No. 502080593
Encompass_Preferred
Medchemexpress, HY-100562 100mg Flopropione CAS:2295-58-1 Flopropione is a 5-HT receptor antagonist and also a catechol-o-methyltransferase (COMT) inhibitor. Flopropione also as an antispasmodic agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0593 Supplier Medchemexpress LLC Supplier No. HY100562100MG
Medchemexpress, HY-100562 100mg Flopropione CAS:2295-58-1 Flopropione is a 5-HT receptor antagonist and also a catechol-o-methyltransferase (COMT) inhibitor. Flopropione also as an antispasmodic agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.