Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Florasulam | 145701-23-1 | 99.4% | 250 MG
SDP

Catalog No. 5000414235
Encompass_Preferred

Florasulam is a targeted post-emergent herbicide belonging to the triazolopyrimidine sulfonanilide class. It functions by inhibiting acetolactate synthase (ALS) in plants, which disrupts plant cell division, reduces growth, and ultimately leads to plant death. ALS is crucial for the biosynthesis of branched-chain amino acids and is located in the chloroplasts.

  • Purity is 99.42%.
  • CAS number is 145701-23-1.
  • Molecular weight is 359.28.
  • Formula is C12H8F3N5O3S.
  • Appearance is solid.
  • Color is white to off-white.
  • Shipping is at room temperature in continental US; may vary elsewhere.
  • Storage is at room temperature for 3 years; in solvent, -80°C for 2 years, -20°C for 1 year.

Catalog No. 50-004-14235 Supplier Medchemexpress LLC Supplier No. 250MGHYB1845
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Florasulam is a targeted post-emergent herbicide belonging to the triazolopyrimidine sulfonanilide class. It functions by inhibiting acetolactate synthase (ALS) in plants, which disrupts plant cell division, reduces growth, and ultimately leads to plant death. ALS is crucial for the biosynthesis of branched-chain amino acids and is located in the chloroplasts.

  • Purity is 99.42%.
  • CAS number is 145701-23-1.
  • Molecular weight is 359.28.
  • Formula is C12H8F3N5O3S.
  • Appearance is solid.
  • Color is white to off-white.
  • Shipping is at room temperature in continental US; may vary elsewhere.
  • Storage is at room temperature for 3 years; in solvent, -80°C for 2 years, -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.