Learn More
Medchemexpress LLC Florbetapir (18F)-radiolabeled styrylpyridine tosylate precursor | 1205550-99-7 | 99.7% | 100 MG
SDP
Catalog No. 5000232035
Encompass_Preferred
A styrylpyridine tosylate precursor used to synthesize 18F-radiolabeled florbetapir-type tracers for positron emission tomography (PET) research.
  • High chemical purity suitable for radiolabeling workflows.
  • Used to synthesize 18F-labeled PET imaging agents targeting amyloid plaques.
  • White to yellow solid; easy handling and storage as a dry powder.
  • Long-term stability when stored at -20°C; solvent solutions require colder storage.
  • Available in small research-scale quantities for tracer development.
Catalog No. 50-002-32035 Supplier Medchemexpress LLC Supplier No. HY120947100MG
A styrylpyridine tosylate precursor used to synthesize 18F-radiolabeled florbetapir-type tracers for positron emission tomography (PET) research.
  • High chemical purity suitable for radiolabeling workflows.
  • Used to synthesize 18F-labeled PET imaging agents targeting amyloid plaques.
  • White to yellow solid; easy handling and storage as a dry powder.
  • Long-term stability when stored at -20°C; solvent solutions require colder storage.
  • Available in small research-scale quantities for tracer development.