Learn More
Medchemexpress LLC Florbetapir (Standard) (AV-45 (Standard)) | 938435-69-9 | 99.5% | 1 MG
SDP
Catalog No. 5000340698
Encompass_Preferred
Florbetapir (Standard) (AV-45 (Standard)) is an analytical standard of Flobetapir (HY-129650) intended for research and analytical applications. Flobetapir is a well-tolerated imaging agent that synthesizes Florbetapir (18F), a PET scanning radiopharmaceutical compound.
  • Used for research and analytical applications
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
  • Molecular weight is 360.42
  • Appears as a solid, light brown to yellow in color
Catalog No. 50-003-40698 Supplier Medchemexpress LLC Supplier No. HY129650R1MG
Florbetapir (Standard) (AV-45 (Standard)) is an analytical standard of Flobetapir (HY-129650) intended for research and analytical applications. Flobetapir is a well-tolerated imaging agent that synthesizes Florbetapir (18F), a PET scanning radiopharmaceutical compound.
  • Used for research and analytical applications
  • Commonly used in qualitative, quantitative and methodological research experiments
  • Suitable for HPLC, GC and MS applications
  • Molecular weight is 360.42
  • Appears as a solid, light brown to yellow in color