Learn More
ELITechGroup Aerospray TB Reagent A, Fluorescence Decolorizer, 500mL
SDP
Catalog No. NC9388773
Encompass
This item has a minimum order quantity of 3 per supplier requirements. Decolorizer; Fluorescence; Acid Fast Reagent AF; Clear liquid with an alcoholic odor; 500mL; B.P. 70 deg. to 90 deg.C; Freezing Point
Catalog No. NC9388773 Supplier ELITechGroup Supplier No. SS061AF
This item has a minimum order quantity of 3 per supplier requirements. Decolorizer; Fluorescence; Acid Fast Reagent AF; Clear liquid with an alcoholic odor; 500mL; B.P. 70 deg. to 90 deg.C; Freezing Point