Learn More
Medchemexpress LLC Florfenicol | 73231-34-2 | 99.7% | 25 MG
SDP
Catalog No. 5000414099
Encompass_Preferred
Florfenicol (Standard) is an analytical standard of Florfenicol, an orally active broad-spectrum antibacterial antibiotic. It exhibits anti-inflammatory, pro-apoptotic, and immunomodulatory functions, making it suitable for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Suitable for laboratory chemical uses and substance manufacturing
  • Intended for research use only
Catalog No. 50-004-14099 Supplier Medchemexpress LLC Supplier No. 25MGHYB1374R
Florfenicol (Standard) is an analytical standard of Florfenicol, an orally active broad-spectrum antibacterial antibiotic. It exhibits anti-inflammatory, pro-apoptotic, and immunomodulatory functions, making it suitable for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Suitable for laboratory chemical uses and substance manufacturing
  • Intended for research use only