Learn More
Medchemexpress LLC Florfenicol amine-d3 | 3023974-39-9 | >=98.0% | 250.30 g/mol | C10H11D3FNO3S | 1 MG
SDP
Catalog No. 5000283916
Encompass_Preferred
Florfenicol amine-d3 is a deuterium-labeled (d3) analogue of florfenicol amine supplied as a laboratory reference standard for analytical testing. It is intended for research use and for quantitative method development using mass spectrometry.
  • Deuterium-labeled internal standard for florfenicol amine.
  • Intended for research use only; not for human or clinical applications.
  • Suitable for quantitative mass spectrometry and method development.
  • High isotopic enrichment (d3) to improve analytical accuracy.
  • Supplied as a solid reference material; handle according to the safety data sheet.
  • Available in 1 mg, 5 mg, 10 mg packages.
Catalog No. 50-002-83916 Supplier Medchemexpress LLC Supplier No. HYW6538711MG
Florfenicol amine-d3 is a deuterium-labeled (d3) analogue of florfenicol amine supplied as a laboratory reference standard for analytical testing. It is intended for research use and for quantitative method development using mass spectrometry.
  • Deuterium-labeled internal standard for florfenicol amine.
  • Intended for research use only; not for human or clinical applications.
  • Suitable for quantitative mass spectrometry and method development.
  • High isotopic enrichment (d3) to improve analytical accuracy.
  • Supplied as a solid reference material; handle according to the safety data sheet.
  • Available in 1 mg, 5 mg, 10 mg packages.