Learn More
Medchemexpress LLC Florfenicol amine (hydrochloride) | 108656-33-3 | 98.6% | 283.75 g/mol | C10H15ClFNO3S | 1 ML
SDP
Catalog No. 5000227828
Encompass_Preferred
Florfenicol amine hydrochloride is the hydrochloride salt of the primary metabolite of the veterinary antibiotic florfenicol. It is provided for research and analytical applications and is available as a ready-to-use 10 mM solution in DMSO (1 mL) or as a solid for reconstitution.
  • High purity material verified by LCMS and COA.
  • Supplied as a pre-made 10 mM solution in DMSO for immediate use.
  • Also available as solid quantities for custom preparation.
  • Suitable as an analytical reference standard and metabolite marker.
  • Characterized by molecular formula C10H15ClFNO3S and molecular weight 283.75 g/mol.
  • Recommended solvent: DMSO for dissolution and handling.
Catalog No. 50-002-27828 Supplier Medchemexpress LLC Supplier No. HY11809910MM 1ML
Florfenicol amine hydrochloride is the hydrochloride salt of the primary metabolite of the veterinary antibiotic florfenicol. It is provided for research and analytical applications and is available as a ready-to-use 10 mM solution in DMSO (1 mL) or as a solid for reconstitution.
  • High purity material verified by LCMS and COA.
  • Supplied as a pre-made 10 mM solution in DMSO for immediate use.
  • Also available as solid quantities for custom preparation.
  • Suitable as an analytical reference standard and metabolite marker.
  • Characterized by molecular formula C10H15ClFNO3S and molecular weight 283.75 g/mol.
  • Recommended solvent: DMSO for dissolution and handling.