Learn More
Medchemexpress LLC Florfenicol amine hydrochloride | 108656-33-3 | 98.6% | 283.75 g/mol | C10H15ClFNO3S | 5 MG
SDP
Catalog No. 5000226827
Encompass_Preferred
Florfenicol amine hydrochloride is the hydrochloride salt of the primary metabolite of florfenicol. It is supplied as a research-grade analytical standard for metabolite studies, residue analysis, and microbiology research.
  • High purity (98.6%) analytical standard.
  • Available as small solid quantities suitable for analytical workflows.
  • Molecular formula C10H15ClFNO3S; molecular weight 283.75 g/mol.
  • Stable when stored sealed and away from moisture; follow solvent storage recommendations for long-term stability.
  • Intended for research use as a metabolite reference and analytical standard.
Catalog No. 50-002-26827 Supplier Medchemexpress LLC Supplier No. HY1180995MG
Florfenicol amine hydrochloride is the hydrochloride salt of the primary metabolite of florfenicol. It is supplied as a research-grade analytical standard for metabolite studies, residue analysis, and microbiology research.
  • High purity (98.6%) analytical standard.
  • Available as small solid quantities suitable for analytical workflows.
  • Molecular formula C10H15ClFNO3S; molecular weight 283.75 g/mol.
  • Stable when stored sealed and away from moisture; follow solvent storage recommendations for long-term stability.
  • Intended for research use as a metabolite reference and analytical standard.