Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Florfenicol amine hydrochloride | 108656-33-3 | MFCD07369400 | >=95.0% | 283.75 g/mol | C10H15ClFNO3S | 10 MG
SDP

Catalog No. 5000307715
Encompass_Preferred

Florfenicol amine hydrochloride is the hydrochloride salt of florfenicol amine, a metabolite of the antibiotic florfenicol. It is provided as a research-grade reagent for analytical reference, metabolite profiling, and in vitro studies of bacterial protein synthesis.

  • Suitable as an analytical standard and metabolite reference.
  • Used in studies of bacterial protein synthesis and antibiotic metabolism.
  • Molecular weight 283.75 g/mol; formula C10H15ClFNO3S.
  • Available in small pack sizes appropriate for research use, e.g., 10 MG.
  • Storage: store sealed at 4°C; in solution, store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-07715 Supplier Medchemexpress LLC Supplier No. HY11809910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Florfenicol amine hydrochloride is the hydrochloride salt of florfenicol amine, a metabolite of the antibiotic florfenicol. It is provided as a research-grade reagent for analytical reference, metabolite profiling, and in vitro studies of bacterial protein synthesis.

  • Suitable as an analytical standard and metabolite reference.
  • Used in studies of bacterial protein synthesis and antibiotic metabolism.
  • Molecular weight 283.75 g/mol; formula C10H15ClFNO3S.
  • Available in small pack sizes appropriate for research use, e.g., 10 MG.
  • Storage: store sealed at 4°C; in solution, store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.