Learn More
Cayman Chemical Florfenicol-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2213400-85-0, 1 mg
SDP
Catalog No. 502938741
Encompass_Preferred
An internal standard for the quantification of florfenicol by GC- or LC-MS
Catalog No. 50-293-8741 Supplier Cayman Chemical Supplier No. 285381 MG
An internal standard for the quantification of florfenicol by GC- or LC-MS