Learn More
Medchemexpress LLC Florfenicol-d3 | 2213400-85-0 | 1MG
SDP
Catalog No. 5000413078
Encompass_Preferred
Florfenicol-d3 is a deuterium-labeled version of Florfenicol, a veterinary antibiotic used for bovine respiratory disease and to control enteric septicemia in catfish. It has been shown to induce early embryonic death in eggs. Florfenicol-d3 can be used as a tracer and as an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, is explored for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has a molecular weight of 361.23.
  • Appears as a white to off-white solid.
Catalog No. 50-004-13078 Supplier Medchemexpress LLC Supplier No. HYB1374S11MG
Florfenicol-d3 is a deuterium-labeled version of Florfenicol, a veterinary antibiotic used for bovine respiratory disease and to control enteric septicemia in catfish. It has been shown to induce early embryonic death in eggs. Florfenicol-d3 can be used as a tracer and as an internal standard for quantitative analysis. Deuteration, the incorporation of stable heavy isotopes, is explored for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Has a molecular weight of 361.23.
  • Appears as a white to off-white solid.