Learn More
Medchemexpress LLC Florfenicol-d3 | 2213400-85-0 | 99.9% | 5 MG
SDP
Catalog No. 5000412883
Encompass_Preferred
Florfenicol-d3 is the deuterium labeled form of Florfenicol, a veterinary antibiotic. It is used for bovine respiratory disease and in aquaculture to control enteric septicemia in catfish. Florfenicol can induce early embryonic death in eggs, with an LC50 of 1.07 μg/g. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-12883 Supplier Medchemexpress LLC Supplier No. HYB1374S15MG
Florfenicol-d3 is the deuterium labeled form of Florfenicol, a veterinary antibiotic. It is used for bovine respiratory disease and in aquaculture to control enteric septicemia in catfish. Florfenicol can induce early embryonic death in eggs, with an LC50 of 1.07 μg/g. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.