Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Florfenicol, G-Biosciences
SCI_ED

Catalog No. S139772
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139772 5 g
S139730 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139772 Supplier G-Biosciences Supplier No. RC1022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Florfenicol, like chloramphenicol, is a bacteriostatic antibiotic and an analog of thiamphenicol.

It targets gram-negative and gram-positive bacteria, especially those responsible for bovine respiratory infections. Florfenicol is soluble in water and lipid solvents.

Chemical Identifiers

Molecular Weight (g/mol) 358.21

Specifications

Quantity 5 g
Product Line Gibco
Boiling Point 618°C
Percent Purity >98%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.