Learn More
SPEX CERTIPREP LLC 3P FLORIDE 250ML 1000PPM
Catalog No. NC9636382
Encompass_Preferred
(HPS by Zeptometrix) 1000 ug/mL Fluoride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9636382 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICFFM250
(HPS by Zeptometrix) 1000 ug/mL Fluoride in H2O; Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.