Learn More
Florisil™ Standard, Sold by MilliporeSigma™ Supelco™
Catalog No. 111022445 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
454 g
Catalog No. Quantity
11-102-2445 454 g
1 options
Catalog No. 11-102-2445 Supplier MilliporeSigma Supelco Supplier No. 20281
Add to Cart
Add to Cart
Florisil, a commercially-prepared magnesia silica gel, for the separation of lipids. It is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.
  • Coarse mesh size provides a rapid flow rate
TRUSTED_SUSTAINABILITY
Specifications
Quantity 454 g
Florisil is a registered trademark of U.S. Silica Company