Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Florsheim Work Boot, Size 9, Leather, Steel Toe, 1 ea

Catalog No. 5000688805
Encompass

This men's work boot is designed for durability and protection, featuring a steel toe and electrical hazard protection. It includes a Foot Force F2A athletic footbed for enhanced comfort.

  • Footwear type: Work boot
  • Gender: Men
  • Men's size: 9
  • Upper material: Leather
  • Outsole material: Polyurethane, rubber
  • Primary color: Black
  • Toe type: Steel
  • Closure type: Slip-on
  • Series: Hercules
  • Shoe width: Medium
  • Provides ASTM F2413 compliance and metatarsal guard
  • Boot height: 6 in.

Catalog No. 50-006-88805 Supplier MSC Supplier No. 18266247
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This men's work boot is designed for durability and protection, featuring a steel toe and electrical hazard protection. It includes a Foot Force F2A athletic footbed for enhanced comfort.

  • Footwear type: Work boot
  • Gender: Men
  • Men's size: 9
  • Upper material: Leather
  • Outsole material: Polyurethane, rubber
  • Primary color: Black
  • Toe type: Steel
  • Closure type: Slip-on
  • Series: Hercules
  • Shoe width: Medium
  • Provides ASTM F2413 compliance and metatarsal guard
  • Boot height: 6 in.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.