Learn More
Aviva Systems Biology Corp FLOT1 Antibody - C-terminal region (ARP65758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FLOT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human FLOT1. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-288-563