Learn More
Electron Microscopy Sciences Flotation Workstation 2 x 6" Slide Dryer, 8 x 8 x 2.25" Dish, 120 VAC
SDP
Catalog No. 501902231
Encompass_Preferred
This Flotation Workstation makes tissue preparation easier, safer and more efficient. Comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled heating, ambient to 65 degree C and a high contrast background for optimum specimen viewing combined with the HISTO Orientator for flattening sections and a Slide Dryer.
Dish size: 8 x 8 x 2.25"
Catalog No. 50-190-2231 Supplier Electron Microscopy Sciences Supplier No. 6321410
This Flotation Workstation makes tissue preparation easier, safer and more efficient. Comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled heating, ambient to 65 degree C and a high contrast background for optimum specimen viewing combined with the HISTO Orientator for flattening sections and a Slide Dryer.
Dish size: 8 x 8 x 2.25"