Learn More
Electron Microscopy Sciences Flotation Workstation 2 x 6" Slide Dryer
SDP
Catalog No. 501902233
Encompass_Preferred
This Flotation Workstation makes tissue preparation easier, safer and more efficient. Comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled heating, ambient to 65 degree C and a high contrast background for optimum specimen viewing combined with the HISTO Orientator for flattening sections and a Slide Dryer.
Dish size: 8 x 8 x 2.25"
Catalog No. 50-190-2233 Supplier Electron Microscopy Sciences Supplier No. 6321420
This Flotation Workstation makes tissue preparation easier, safer and more efficient. Comes with conveniently removable glass dish, scratch and chemical resistant plastic housing, microprocessor controlled heating, ambient to 65 degree C and a high contrast background for optimum specimen viewing combined with the HISTO Orientator for flattening sections and a Slide Dryer.
Dish size: 8 x 8 x 2.25"