Learn More
Enzo Life Sciences FLUOR DE LYS deacetylase substrate (50 µl)
SDP
Supplier:  Enzo Life Sciences BMLKI1040050
Encompass_Preferred
Highly sensitive and convenient alternative to radiolabeled, acetylated histones or peptide/HPLC methods for the assay of histone deacetylases. MW: 387.4. Formulation: Supplied as a 50 mM solution (50 µl) in dimethylsulfoxide (DMSO). Purity: ≥95% (HPLC). Long Term Storage: -80°C
Catalog No. NC9915672