Learn More
Starna Cells Inc FLOURES CELL W/STOPPER MICRO
SDP
Catalog No. NC9163538
Encompass_Preferred
Window material Spectrosil Quartz or equivalent, useable range: 170 to 2700nm, Micro Fluorometer Cells, with Stopper Type 28F, Physical Configuration: Thick windows parallel to light path, sample chamber 2mm wide
Catalog No. NC9163538 Supplier Starna Cells Inc Supplier No. 28FQ10
Window material Spectrosil Quartz or equivalent, useable range: 170 to 2700nm, Micro Fluorometer Cells, with Stopper Type 28F, Physical Configuration: Thick windows parallel to light path, sample chamber 2mm wide