Learn More
Starna Cells Inc FLOURESENCE CELL W/STOPPER
SDP
Catalog No. NC9973006
Encompass_Preferred
Semi-Micro Fluorometer Cells,with Stopper,Window material: Spectrosil Quartz or equivalent,useable range:170 to 2700 nm, Path Length:10 mm, Exterior(W x L x H):12.5 x 12.5 x 48 mm, Interior (W x L):4 x 10 mm, Nominal Volume :1.400mL
Catalog No. NC9973006 Supplier Starna Cells Inc Supplier No. 29FQ10
Semi-Micro Fluorometer Cells,with Stopper,Window material: Spectrosil Quartz or equivalent,useable range:170 to 2700 nm, Path Length:10 mm, Exterior(W x L x H):12.5 x 12.5 x 48 mm, Interior (W x L):4 x 10 mm, Nominal Volume :1.400mL