Learn More
Starna Cells Inc FLOUROMETER 10MM QUARTZ
SDP
Catalog No. NC9781397
Encompass_Preferred
Standard Fluorometer Cells, with Stopper,Window material:Spectrosil Quartz or equivalent,useable range: 170 to 2700nm, Path Length:10 mm, Exterior (W x L x H):12.5 x 12.5 x 48 mm, Interior (W x L):10 x 10 mm, Nominal Volume :3.500ml, Number
Catalog No. NC9781397 Supplier Starna Cells Inc Supplier No. 23Q10
Standard Fluorometer Cells, with Stopper,Window material:Spectrosil Quartz or equivalent,useable range: 170 to 2700nm, Path Length:10 mm, Exterior (W x L x H):12.5 x 12.5 x 48 mm, Interior (W x L):10 x 10 mm, Nominal Volume :3.500ml, Number