Learn More
FEEGER LUCAS WOLFE BELLOFRAM TYPE 70 BP HIGH FLOW BACK PRESSURE AIR REGULATOR, 1/4" NPT, 0-2 PSI, 960-191-000
Catalog No. NC2737983
Encompass
The Type 70BP High-Flow Back Pressure Air Regulator is a high-flow, high-precision pneumatic relief valve with adjustable set point. It is expressly designed for applications requiring high flow capacity and accurate pressure controls. It may be used in place of a standard relief valve to improve pressure control while maintaining protection against over-pressurization of supply pressure lines to instruments and other control devices.The Type 70BP is most often used in conjunction with diaphragm-operated valve and bleed orifice as a compressor controller. Excess compressor pressure is relieved through the regulator to actuate the intake restricting valve. When the regulator is shut off, downstream air escapes through the bleed orifice, allowing the restricting valve to open. Units are designed to respond quickly to minute changes in upstream pressure. Incorporation of a patented Bellofram rolling diaphragm design offers reliable perfo
Catalog No. NC2737983 Supplier FEEGER LUCAS WOLFE Supplier No. 960191000
The Type 70BP High-Flow Back Pressure Air Regulator is a high-flow, high-precision pneumatic relief valve with adjustable set point. It is expressly designed for applications requiring high flow capacity and accurate pressure controls. It may be used in place of a standard relief valve to improve pressure control while maintaining protection against over-pressurization of supply pressure lines to instruments and other control devices.The Type 70BP is most often used in conjunction with diaphragm-operated valve and bleed orifice as a compressor controller. Excess compressor pressure is relieved through the regulator to actuate the intake restricting valve. When the regulator is shut off, downstream air escapes through the bleed orifice, allowing the restricting valve to open. Units are designed to respond quickly to minute changes in upstream pressure. Incorporation of a patented Bellofram rolling diaphragm design offers reliable perfo