Learn More
BUHLMANN DIAGNOSTICS FLOW CAST BASOPHIL ACTIVATION
Catalog No. NC1744271
Encompass
  • Comprehensive allergy detection with capability for IgE and non-IgE mediated reactions.
  • Simplified basophil identification through dual labeling with CCR3-PE and CD63-FITC.
  • Enhanced precision and reliability using specific FcεRI receptor agonization and validated controls.
  • Optimized workflow with sample stability up to 5 days when fixed and stored at 2-8°C.
  • Compatible with a wide range of BÜHLMANN allergens.
  • Regulatory compliance as RUO in the US and IVDR cleared in other regions.
  • Uses whole blood, EDTA samples for testing.
  • Includes positive and negative controls for accurate results.
  • Storage capability for whole blood samples up to 48 hours at 2-8°C.
  • Minimum stimulation index of 2 for a positive result.
  • Requires sterile, ultrapure, apyrogenic water.
Catalog No. NC1744271 Supplier BUHLMANN DIAGNOSTICS Supplier No. FKCCRU
  • Comprehensive allergy detection with capability for IgE and non-IgE mediated reactions.
  • Simplified basophil identification through dual labeling with CCR3-PE and CD63-FITC.
  • Enhanced precision and reliability using specific FcεRI receptor agonization and validated controls.
  • Optimized workflow with sample stability up to 5 days when fixed and stored at 2-8°C.
  • Compatible with a wide range of BÜHLMANN allergens.
  • Regulatory compliance as RUO in the US and IVDR cleared in other regions.
  • Uses whole blood, EDTA samples for testing.
  • Includes positive and negative controls for accurate results.
  • Storage capability for whole blood samples up to 48 hours at 2-8°C.
  • Minimum stimulation index of 2 for a positive result.
  • Requires sterile, ultrapure, apyrogenic water.