Learn More
Azzota Corp Flow cell 10mm 0.48ml Qz
Catalog No. 5000887024
Encompass_Preferred
Self masking continuous flowthrough cell 10mm Pathlength window size 4x12mm 0 48ml Quartz 190-2500nm
Catalog No. 50-008-87024 Supplier Azzota Corp Supplier No. Q74
Self masking continuous flowthrough cell 10mm Pathlength window size 4x12mm 0 48ml Quartz 190-2500nm