Learn More
Waters Corp Waters Corporation Flow cell, 3 mm Copper WE, Hy-REF Reference
Catalog No. 50791703 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This flow cell kit features a 3 mm Copper Working Electrode and a Hy-REF Reference. It is suitable for carbohydrate analysis and high pH mobile phases. The Hy-REF is maintenance-free, and the overall design is rugged and flexible, allowing for electrode customization and easy maintenance.
  • Features a 3 mm copper working electrode and a Hy-REF reference.
  • Hy-REF reference electrode is ideal for carbohydrate analysis.
  • Compatible with high pH mobile phases.
  • Entirely maintenance-free Hy-REF reference electrode.
  • Rugged flow cell design ensures maximum system uptime.
  • Design allows users to mix-and-match working and reference electrodes.
  • Lowers total cost-of-ownership.
  • Enhances overall system flexibility.
  • Easy to install.
  • Easy to re-assemble after cleaning or maintenance.
Catalog No. 50-791-703 Supplier Waters Corp Supplier No. 205004335
Add to Cart
Add to Cart
Request a Quote
This flow cell kit features a 3 mm Copper Working Electrode and a Hy-REF Reference. It is suitable for carbohydrate analysis and high pH mobile phases. The Hy-REF is maintenance-free, and the overall design is rugged and flexible, allowing for electrode customization and easy maintenance.
  • Features a 3 mm copper working electrode and a Hy-REF reference.
  • Hy-REF reference electrode is ideal for carbohydrate analysis.
  • Compatible with high pH mobile phases.
  • Entirely maintenance-free Hy-REF reference electrode.
  • Rugged flow cell design ensures maximum system uptime.
  • Design allows users to mix-and-match working and reference electrodes.
  • Lowers total cost-of-ownership.
  • Enhances overall system flexibility.
  • Easy to install.
  • Easy to re-assemble after cleaning or maintenance.