Learn More
Anton Paar Micro Flow Cell for Abbemat 300/350/500/550
Catalog No. NC1818555
Encompass_Preferred
This type of flow cell is used for continuous measurements of liquids. The micro flow cell offers advantages when combined with an automated system such as a peristaltic pump.Suitable for use with Anton Paar refractometersAbbemat 300 item NC1818534,Abbemat 350 item NC1818535,Abbemat 500 item NC1818537,Abbemat 550 item NC1818539
Catalog No. NC1818555 Supplier Anton Paar Supplier No. 101384
This type of flow cell is used for continuous measurements of liquids. The micro flow cell offers advantages when combined with an automated system such as a peristaltic pump.Suitable for use with Anton Paar refractometersAbbemat 300 item NC1818534,Abbemat 350 item NC1818535,Abbemat 500 item NC1818537,Abbemat 550 item NC1818539