Learn More
Azzota Corp Flow cell 5mm 0.24ml Qz
Catalog No. 5000887025
Encompass_Preferred
Self masking continuous flowthrough cell 5mm Pathlength window size 4x6mm 0 24ml Quartz 190-2500nm
Catalog No. 50-008-87025 Supplier Azzota Corp Supplier No. Q73
Self masking continuous flowthrough cell 5mm Pathlength window size 4x6mm 0 24ml Quartz 190-2500nm