Learn More
Waters Corp Waters Flow Cell Kit, 2 mm Glassy Carbon Working Electrode, Salt-Bridge Reference
Catalog No. 50791698 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This flow cell kit is a critical component for electrochemical detection systems. It features a 2 mm glassy carbon working electrode and a salt-bridge reference, providing reliable performance in various laboratory applications.
  • Features a 2 mm glassy carbon working electrode.
  • Includes a salt-bridge reference.
  • Designed as a flow cell option for electrochemical detectors.
  • Compatible with the 2465 Electrochemical Detector.
  • Composed of a working electrode block and an inlet (reference) block, joined by bolts.
Catalog No. 50-791-698 Supplier Waters Corp Supplier No. 205004115
Add to Cart
Add to Cart
Request a Quote
This flow cell kit is a critical component for electrochemical detection systems. It features a 2 mm glassy carbon working electrode and a salt-bridge reference, providing reliable performance in various laboratory applications.
  • Features a 2 mm glassy carbon working electrode.
  • Includes a salt-bridge reference.
  • Designed as a flow cell option for electrochemical detectors.
  • Compatible with the 2465 Electrochemical Detector.
  • Composed of a working electrode block and an inlet (reference) block, joined by bolts.