Learn More
Thermo Scientific Vanquish Variable Wavelength - Semi-Micro Flow Cell | | 1 EA
Catalog No. NC1917092 Shop All Thermo Scientific Products
Encompass
For Use With Equipment: Vanquish Variable Wavelength Detector F, Material: Stainless Steel, Product Line: Vanquish, Type: Semi-Micro Flow Cell, Volume Metric: 25 L
This item is not returnable. View return policy
Catalog No. NC1917092 Supplier Thermo Scientific Supplier No. 6077.0360
For Use With Equipment: Vanquish Variable Wavelength Detector F, Material: Stainless Steel, Product Line: Vanquish, Type: Semi-Micro Flow Cell, Volume Metric: 25 L