Learn More
Quanterix Inc FLOW CELLS FOR PHENOCYCLER-FUS
SDP
Catalog No. NC2338026
Encompass
  • Flow cells enable ultrahigh-plex spatial phenotyping and multiplex biomarker analysis.
  • Compatible with the PhenoCycler-Fusion system for optimal performance.
  • Facilitates single-cell resolution analysis on whole microscope slides.
  • Designed for seamless integration with automated fluidics.
  • Enables comprehensive biological insights by interrogating biomarkers in spatial context.
  • Closed system design reduces contamination risks and user error.
  • Ensures consistent and high-quality experimental outcomes.
  • Supports 10 individual experiments for enhanced workflow efficiency.
  • Shipping condition is ambient for straightforward storage and handling.
  • Suitable for spatial phenotyping, multiplex biomarker analysis, and tissue analysis.
Catalog No. NC2338026 Supplier Quanterix Inc Supplier No. 240205
  • Flow cells enable ultrahigh-plex spatial phenotyping and multiplex biomarker analysis.
  • Compatible with the PhenoCycler-Fusion system for optimal performance.
  • Facilitates single-cell resolution analysis on whole microscope slides.
  • Designed for seamless integration with automated fluidics.
  • Enables comprehensive biological insights by interrogating biomarkers in spatial context.
  • Closed system design reduces contamination risks and user error.
  • Ensures consistent and high-quality experimental outcomes.
  • Supports 10 individual experiments for enhanced workflow efficiency.
  • Shipping condition is ambient for straightforward storage and handling.
  • Suitable for spatial phenotyping, multiplex biomarker analysis, and tissue analysis.